Z-Hyp(tBu)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Z-Hyp(Tbu)-OH, trans-N-Benzyloxycarbonyl-4-tert-butoxy-L-proline, Cbz-Hyp(tBu)-OH
4008169.0005
4008169.0025
C-3150.0005BAEA
261.76
USD
C-3150.0005BA
C-3150.0025BA
Z-Hyp(tBu)-OH
Z-Hyp(tBu)-OH
Sequence: Z-Hyp(tBu)-OH
Hyp denotes L-trans-hydroxyproline.
Formula:
C₁₇H₂₃NO₅ MW: 321.373 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00153336 CAS Number: 85201-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...