Z-D-Orn(Boc)-OH
Supplier: BACHEM AMERICAS INC
4011139.0005
4011139.0025
C-3070.0005BAEA
653.59
USD
C-3070.0005BA
C-3070.0025BA
Z-D-Orn(Boc)-OH
Z-D-Orn(Boc)-OH
Sequence: Z-D-Orn(Boc)-OH
Formula:
C₁₈H₂₆N₂O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00076971 CAS Number: 98264-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...