N-Carbobenzoxy-L-threonine benzyl ester
Supplier: BACHEM AMERICAS INC
Synonyms:
Z-Thr-OBzl, N-Cbz-L-threonine Benzyl Ester
4003488.0005
4003488.0025
C-2640.0005BAEA
133.97
USD
C-2640.0005BA
C-2640.0025BA
N-Carbobenzoxy-L-threonine benzyl ester
N-Carbobenzoxy-L-threonine benzyl ester
Sequence: Z-Thr-OBzl
Bachem also offers phosphothreonine derivatives.
Formula:
C₁₉H₂₁NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00077041 CAS Number: 16597-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...