Z-Sar-OSu

Supplier: BACHEM AMERICAS INC

4008051.0005 4008051.0025
C-2575.0005BAEA 234.5 USD
C-2575.0005BA C-2575.0025BA
Z-Sar-OSu
Z-Sar-OSu

Sequence: Z-Sar-OSu


Bachem also offers Fmoc-Sar-Wang resin (D-1805) and Fmoc-Sar-SASRIN resin (-D-1645).


Synonym(s): Z-N-Me-Gly-OSu

Formula: C₁₅H₁₆N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00153340
CAS Number: 53733-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR