N-Carbobenzoxy-D-glutamic acid

Supplier: BACHEM AMERICAS INC

Synonyms: N-Benzyloxycarbonyl-D-glutamic acid, N-Cbz-D-glutamic Acid, Z-D-Glu-OH, Cbz-D-Glu-OH

4001810.0005 4001810.0100
C-1565.0005BAEA 111.18 USD
C-1565.0005BA C-1565.0100BA
N-Carbobenzoxy-D-glutamic acid
N-Carbobenzoxy-D-glutamic acid

Sequence: Z-D-Glu-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₃H₁₅NO₆
MW: 281.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063193
CAS Number: 63648-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR