Boc-Thr(Leu-Fmoc)-OH
Supplier: BACHEM AMERICAS INC
4061392.0001 4061392.0005
B-3975.0001BAEA 350.02 USD
B-3975.0001BA B-3975.0005BA
Boc-Thr(Leu-Fmoc)-OH
Proteins and Peptides
Isoacyl dipeptides have been developed as building blocks for preventing aggregation of the growing peptide chains during Fmoc-SPPS. As the isopeptide structure is not affected by the final acidolytic cleavage the solubility of the crude product is improved facilitating the subsequent purification. The depsipeptide will rearrange yielding the desired product in slightly basic solution.