Fmoc-Trp(ME)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-methyl-1H-indol-3-yl)propanoic acid
4052226.0001
4052226.0005
B-3875.0001BAEA
334.97
USD
B-3875.0001BA
B-3875.0005BA
Fmoc-Trp(ME)-OH
Fmoc-Trp(ME)-OH
Sequence: Fmoc-Trp(Me)-OH
MeTrp, αMeTrp.
Fmoc-Trp(Me) was employed in the Fmoc-SPPS of compstatin analogs.
Formula:
C₂₇H₂₄N₂O₄ MW: 440.499 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD02684512 CAS Number: 1334509-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...