Fmoc-D-allo-thr(tBu)-OH

Supplier: BACHEM AMERICAS INC

4033844.0001 4033844.0005
B-3700.0001BAEA 1062.08 USD
B-3700.0001BA B-3700.0005BA
Fmoc-D-allo-thr(tBu)-OH
Fmoc-D-allo-thr(tBu)-OH

Sequence: Fmoc-D-allo-Thr(tBu)-OH


Bachem also offers phosphothreonine derivatives.
aThr derivatives.

Formula: C₂₃H₂₇NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00077074
CAS Number: 170643-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR