(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclopentylacetic acid

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Cpg-OH, N-Fmoc-2-cyclopentyl-L-glycine

4040958.0250 4040958.1000
B-3640.0250BAEA 361.05 USD
B-3640.0250BA B-3640.1000BA
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclopentylacetic acid
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclopentylacetic acid

Sequence: Fmoc-Cpg-OH


Bachem offers derivatives of α-aminoglycine, cyclohexylglycine, cyclopentylglycine, and the PNA building block N-β-aminoethylglycine. For t-butylglycine derivatives see t-Leucine.
Cpg derivatives.


Synonym(s): Fmoc-cyclopentyl-Gly-OH#(S)-2-Cyclopentyl-2-(Fmoc-amino)-acetic acid

Formula: C₂₂H₂₃NO₄
MW: 365.429 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01311751
CAS Number: 220497-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR