Fmoc-N-ME-Trp(boc)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-N-Me-Trp(Boc)-OH
4040147.0001
4040147.0005
B-3625.0001BAEA
502.47
USD
B-3625.0001BA
B-3625.0005BA
Fmoc-N-ME-Trp(boc)-OH
Fmoc-N-ME-Trp(boc)-OH
Sequence: Fmoc-N-Me-Trp(Boc)-OH
MeTrp, αMeTrp.
Formula:
C₃₂H₃₂N₂O₆ MW: 540.616 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD02094403 CAS Number: 197632-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...