Fmoc-Tyr(HPO3Bzl)-OH
Supplier: BACHEM AMERICAS INC
Sequence: Fmoc-Tyr(PO(OBzl)OH)-OH
Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
For stable phosphotyrosine mimics see the O-malonyltyrosine derivative B-2825 and the 4-phosphonophenylalanine derivatives B-3895 and A-4725.
Alternative to Fmoc-Tyr(PO₃(MDPSE)₂)-OH.
Formula:
C₃₁H₂₈NO₈P Storage Temperature: Freezer |
MDL Number:
MFCD00797871 CAS Number: 191348-16-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...