Fmoc-N-Me-Thr(tBu)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-(tert-butoxy)butanoic acid, N-Fmoc-O-tert-butyl-N-methyl-L-threonine

4029146.0001 4029146.0005
B-3420.0001BAEA 571.91 USD
B-3420.0001BA B-3420.0005BA
Fmoc-N-Me-Thr(tBu)-OH
Fmoc-N-Me-Thr(tBu)-OH

Sequence: Fmoc-N-Me-Thr(tBu)-OH


Bachem also offers phosphothreonine derivatives.

Formula: C₂₄H₂₉NO₅
MW: 411.498 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02094431
CAS Number: 117106-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR