Fmoc-Cys(Mtt)-OH
Supplier: BACHEM AMERICAS INC
4028614.0005
4028614.0025
B-3340.0005BAEA
163.35
USD
B-3340.0005BA
B-3340.0025BA
Fmoc-Cys(Mtt)-OH
Fmoc-Cys(Mtt)-OH
Sequence: Fmoc-Cys(Mtt)-OH
Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).
Formula:
C₃₈H₃₃NO₄S MW: 599.745 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD02259492 CAS Number: 269067-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...