Fmoc-Phe(4-CN)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-4-cyano-Phe-OH

4027954.0001 4027954.0005
B-3125.0001BAEA 586.19 USD
B-3125.0001BA B-3125.0005BA
Fmoc-Phe(4-CN)-OH
Fmoc-Phe(4-CN)-OH

Sequence: Fmoc-4-cyano-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.


Derivative for introducing the fluorophore p-cyanophenylalanine during Fmoc-SPPS. Educt for the synthesis of Fmoc-p-amidino-phenylalanine, which can be used for introducing an arginine mimetic.

Formula: C₂₅H₂₀N₂O₄
MW: 412.445 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00672559
CAS Number: 173963-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR