Fmoc-L-allo-threoninol
Supplier: BACHEM AMERICAS INC
4027548.0001
4027548.0005
B-3080.0001BAEA
490.19
USD
B-3080.0001BA
B-3080.0005BA
Fmoc-L-allo-threoninol
Fmoc-L-allo-threoninol
Sequence: Fmoc-L-allo-threoninol
Formula:
C₁₉H₂₁NO₄ MW: 327.38 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01318737 CAS Number: 252049-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...