Fmoc-L-allo-isoleucine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-allo-Ile-OH, (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylpentanoic acid

4026721.0001 4026721.0005
B-2880.0001BAEA 524.54 USD
B-2880.0001BA B-2880.0005BA
Fmoc-L-allo-isoleucine
Fmoc-L-allo-isoleucine

Sequence: Fmoc-allo-Ile-OH

Formula: C₂₁H₂₃NO₄
MW: 353.418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631976
CAS Number: 251316-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR