Fmoc-β-(3-benzothienyl)-Ala-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-3-Ala(3-benzothienyl)-OH, Fmoc-β-(3-benzothienyl)-Ala-OH

4026385.0001 4026385.0005
B-2830.0001BAEA 245.01 USD
B-2830.0001BA B-2830.0005BA
Fmoc-β-(3-benzothienyl)-Ala-OH
Fmoc-beta-(3-benzothienyl)-Ala-OH

Sequence: Fmoc-β-(3-benzothienyl)-Ala-OH


3-Benzothienylalanine (3-Bta) is a sulfur analog of tryptophan.

Formula: C₂₆H₂₁NO₄S
Storage Temperature: Freezer
MDL Number: MFCD00672562
CAS Number: 177966-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR