Fmoc-L-Asn(GlcNAc(Ac)3-β)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-Asn(GlcNAc(Ac)₃-β-D)-OH, N-α-(Fluorenylmethoxycarbonyl)-N-γ-(2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl)-L-asparagine, Fmoc-L-Asn((Ac)3-β-D-GlcNAc)-OH
4025504.0100
4025504.0250
B-2480.0100BAEA
770.42
USD
B-2480.0100BA
B-2480.0250BA
Fmoc-L-Asn(GlcNAc(Ac)3-β)-OH
Fmoc-L-Asn(GlcNAc(Ac)3-beta)-OH
Sequence: Fmoc-Asn(GlcNAc(Ac)₃-β-D)-OH
For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".
Synonym(s): N-α-Fmoc-N-γ-(2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-β-D-glucopyranosyl)-L-asparagine
Formula:
C₃₃H₃₇N₃O₁₃ Storage Temperature: Freezer |
MDL Number:
MFCD00237079 CAS Number: 131287-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...