Fmoc-Asn(Mtt)-OH

Supplier: BACHEM AMERICAS INC

4017007.0005 4017007.0025
B-2045.0005BAEA 310.5 USD
B-2045.0005BA B-2045.0025BA
Fmoc-Asn(Mtt)-OH
Fmoc-Asn(Mtt)-OH

Sequence: Fmoc-Asn(Mtt)-OH


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".


Mtt carboxamide protection is superior to Trt when synthesizing peptides containing an N-terminal Asn, as the latter is cleaved only sluggishly in this position.

Formula: C₃₉H₃₄N₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00237160
CAS Number: 144317-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR