Fmoc-N-Me-Phe-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-N-methyl-L-phenylalanine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-phenylpropanoic acid, Fmoc-N-Me-Phe-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-phenylalanine

4014121.0001 4014121.0005
B-1725.0001BAEA 167.52 USD
B-1725.0001BA B-1725.0005BA
Fmoc-N-Me-Phe-OH
Fmoc-N-Me-Phe-OH

Sequence: Fmoc-N-Me-Phe-OH


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
Short N-methyl-phenylalanine-rich peptides were shown to function as versatile blood-brain barrier shuttles.

Formula: C₂₅H₂₃NO₄
MW: 401.462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151938
CAS Number: 77128-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR