Fmoc-Met-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-L-methionine, FMOC-L-Methionine

4003154.0025 4003154.0100 4003154.0500
B-1360.0025BAEA 111.18 USD
B-1360.0025BA B-1360.0100BA B-1360.0500BA
Fmoc-Met-OH
Fmoc-Met-OH

Sequence: Fmoc-Met-OH


Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.

Formula: C₂₀H₂₁NO₄S
MW: 371.46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037134
CAS Number: 71989-28-1
EINECS: 276-258-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR