Fmoc-Gly-Osu

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Gly-OSu

4003175.0001 4003175.0005 4003175.0025
B-1335.0001BAEA 115.88 USD
B-1335.0001BA B-1335.0005BA B-1335.0025BA
Fmoc-Gly-Osu
Fmoc-Gly-Osu

Sequence: Fmoc-Gly-OSu


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Formula: C₂₁H₁₈N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00065651
CAS Number: 113484-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR