Boc-Gly-Gly-Gly-OH
Supplier: BACHEM AMERICAS INC
4003590.0005 4003590.0025
A-4380.0005BAEA 167.34 USD
A-4380.0005BA A-4380.0025BA
Boc-Gly-Gly-Gly-OH
Proteins and Peptides
For Boc-dipeptides Boc-Xaa-Gly-OH and Boc-Xaa-Pro-OH, which can be used as building blocks during Boc-SPPS, please see the product family 'Boc-Dipeptide Building Blocks' in the 'Amino Acid Derivatives' section. This family also includes short protected peptide fragments suitable for solution synthesis and preparation of enzyme substrates and inhibitors.
Boc-triglycine.