Boc-D-Dab(Fmoc)-OH

Supplier: BACHEM AMERICAS INC

4027507.0001 4027507.0005
A-4230.0001BAEA 490.19 USD
A-4230.0001BA A-4230.0005BA
Boc-D-Dab(Fmoc)-OH
Boc-D-Dab(Fmoc)-OH

Sequence: Boc-D-Dab(Fmoc)-OH

Formula: C₂₄H₂₈N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00798625
CAS Number: 131570-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR