Boc-n-Me-abz-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
2-((tert-Butoxycarbonyl)(methyl)amino)benzoic acid, 2-(N-Boc-N-methylamino)benzoic acid
4025576.0001
4025576.0005
A-3715.0001BAEA
196.08
USD
A-3715.0001BA
A-3715.0005BA
Boc-n-Me-abz-OH
Boc-n-Me-abz-OH
Sequence: Boc-N-Me-Abz-OH
Synonym(s): Boc-N-methyl-anthranilic acid#Boc-MANT
Formula:
C₁₃H₁₇NO₄ MW: 251.282 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00237567 CAS Number: 141871-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...