H-DAB(boc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: H-Daba(Boc)-OH

4016008.0001 4016008.0005
A-3305.0001BAEA 234.61 USD
A-3305.0001BA A-3305.0005BA
H-DAB(boc)-OH
H-DAB(boc)-OH

Sequence: H-Dab(Boc)-OH

Formula: C₉H₁₈N₂O₄
MW: 218.253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236842
CAS Number: 10270-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR