Boc-Pro-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid

4000220.0100 4000220.0500
A-2235.0100BAEA 143.85 USD
A-2235.0100BA A-2235.0500BA
Boc-Pro-OH
Boc-Pro-OH

Sequence: Boc-Pro-OH

Formula: C₁₀H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00037324
CAS Number: 15761-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR