Boc-N-methyl-L-serine

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-N-Me-Ser-OH, N-Boc-N-methyl-L-serine

4002217.0001 4002217.0005
A-2085.0001BAEA 130.7 USD
A-2085.0001BA A-2085.0005BA
Boc-N-methyl-L-serine
Boc-N-methyl-L-serine

Sequence: Boc-N-Me-Ser-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₉H₁₇NO₅
MW: 219.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037249
CAS Number: 101772-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR