1-Benzyl-N-(tert-butoxycarbonyl)-L-glutamate

Supplier: BACHEM AMERICAS INC

Synonyms: 1-Benzyl N-Boc-L-glutamate, N-(tert-Butoxycarbonyl)-L-glutamic Acid 1-Benzyl Ester, N-Boc-L-glutamic Acid 1-Benzyl Ester, Boc-Glu-OBzl

4000338.0005 4000338.0025
A-1630.0005BAEA 114.5 USD
A-1630.0005BA A-1630.0025BA
1-Benzyl-N-(tert-butoxycarbonyl)-L-glutamate
1-Benzyl-N-(tert-butoxycarbonyl)-L-glutamate

Sequence: Boc-Glu-OBzl


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₇H₂₃NO₆
MW: 337.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065568
CAS Number: 30924-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR