Boc-Thr(tBu)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
(2S,3R)-3-(tert-Butoxy)-2-((tert-butoxycarbonyl)amino)butanoic acid, N-Boc-O-tert-butyl-L-threonine
4009568.0001
4009568.0005
4009568.0025
A-1440.0001BAEA
114.5
USD
A-1440.0001BA
A-1440.0005BA
A-1440.0025BA
Boc-Thr(tBu)-OH
Boc-Thr(tBu)-OH
Sequence: Boc-Thr(tBu)-OH
Bachem also offers phosphothreonine derivatives.
Formula:
C₁₃H₂₅NO₅ MW: 275.345 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00151115 CAS Number: 13734-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...