Econo-Pac® Chromatography Columns, Empty, Gravity-Flow, Bio-Rad
Supplier: Bio-Rad
7321010 7321011
101646-496EA 333.85 USD
101646-496 101646-498
Econo-Pac® Chromatography Columns, Empty, Gravity-Flow, Bio-Rad
Chromatography Columns
Econo-Pac Columns are empty polypropylene columns for gravity-flow chromatography.
For higher flow rates, to eliminate the head space above the gel, and to protect the top of the bed from disruption during sample loading, use the Econ-Pac Flow Adaptor.
The columns have graduated volume markings, and can be used with media volumes from 1 to 20 ml. Columns come with an upper bed support (frit) for use with open columns to prevent them from running dry. The luer end fitting, with a snap-off tip, permits the attachment of fittings. The columns can be autoclaved.
Información para pedidos: Each column has an upper bed support, end cap, and tip closure. In addition to the Econ-Pac Flow Adaptor, accessories include female luer end plugs, two-way stopcocks, and the Poly Column 20-place rack.