Multiplate™ 96-Well Unskirted PCR Plates, High-Profile, Bio-Rad
Supplier: Bio-Rad
MLP9621 MLP9631 MLP9641 MLP9611 MLP9651 MLP9601
101647-190EA 261.54 USD
101647-190 101647-192 101647-194 101647-188 101647-196 101647-186
Multiplate™ 96-Well Unskirted PCR Plates, High-Profile, Bio-Rad
Microplates PCR Plates
The versatile Multiplate™ 96-Well Unskirted PCR Plates are compatible with many instruments and a wide variety of sealing systems for oil-free cycling
  • 5–125 µl recommended volume range (maximum 300 µl)
  • Available in clear and a range of colors
  • Free of DNase, RNase, and human DNA
These polypropylene PCR plates have very low protein binding and excellent retention of sample volume. When less than a full plate is needed, plates can be easily cut with scissors to the required size.
Compatible with: Bio-Rad™ 1000 series, Bio-Rad DNA Engine® series, Bio-Rad MJ Mini™, Bio-Rad CFX96™, Eppendorf Mastercycler series, Agilent (Stratagene) MX series, Idaho Technology LightScanner, Life Technology DNA sequencers.