Microseal® 96-Well Skirted PCR Plates, Low-Profile, Bio-Rad
Supplier: Bio-Rad
MSP9605 MSP9631 MSP9651 MSP9601
101647-224EA 550.77 USD
101647-224 101647-230 101647-236 101647-222 101647-226 101647-238 101647-232
Microseal® 96-Well Skirted PCR Plates, Low-Profile, Bio-Rad
Microplates PCR Plates
Microseal® 96-Well Skirted PCR Plates, Low Profile, feature low-binding polypropylene construction and a skirted design that is suitable for high-throughput microplate handling including most robotic workstations, microplate readers, and robotic arms.
  • A variety of colors, including opaque white and black for luminescence- and fluorescence-based assays
  • Skirted plates can be gripped by robotic arms
  • Footprint and well spacing meet ANSI/SBS microplate standards
  • Barcoding option available
  • PCR sample volumes of 5–125 µL (220 µL maximum)
  • Free of DNase, RNase, and human DNA
  • Robot compatible
The robot-friendly design features locator holes and flat vertical sidewalls for secure handling and easy barcoding. Raised rims around wells provide an excellent surface for tight sealing with a variety of sealing systems and allow easy release of the sealer from the plate.