Silver Stains, Bio-Rad
Supplier: BIO-RAD LABORATORIES MS
1610472 1610481 1610478 1610448 1610464 1610474 1610473 1610475 1610463 1610479 1610461 1610445 1610477 1610482 1610443 1610462 1610476 1610444 1610449
101643-294EA 167.69 USD
101643-294 101643-312 101643-306 101643-274 101643-288 101643-298 101643-296 101643-300 101643-286 101643-308 101643-282 101643-270 101643-304 101643-314 101643-266 101643-272 101643-284 101643-302 101643-268 101643-276 101643-310
Silver Stains, Bio-Rad
Stains Electrophoresis Stains
The Silver Stain Plus Kit™ is a quick, simple system for detecting proteins or nucleic acids in polyacrylamide and dried agarose gels for 13 full–size or 40 mini gels.
The Silver Stain Plus™ Kit can detect as little as 0.6 ng of protein and DNA, and requires only three steps, allowing visualization of your samples in less than 1 hour. The stain is 30–50 fold more sensitive than Coomassie R250 Dye and 2–5 fold more sensitive than ethidium bromide.