Petri Dishes, PFA, Savillex
Supplier: SAVILLEX
700-100 700-125
30617-524EA 74.51 USD
30617-524 30617-526
Petri Dishes, PFA, Savillex
Culture Dishes Petri Dishes
PFA petri dishes are translucent, chemically inert, and noncontaminating
Service temperature range is –200 to 260°C (–328 to 500°F).
Flat bottomed and stackable. Can also be used as a cover.