Salmon (Des-Gly¹⁰,D-Arg⁶,Pro-NHEt⁹)-LHRH Acetate
Supplier: BACHEM AMERICAS INC
4016385.0005 4016385.0025
H-9205.0005BAEA 490.19 USD
H-9205.0005BA H-9205.0025BA
Salmon (Des-Gly¹⁰,D-Arg⁶,Pro-NHEt⁹)-LHRH Acetate
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Contrary to mammals and other classes of species, the sequences of GnRH I-III found in fish show a high diversity. H-4258, Lamprey GnRH III (peforelin) has found use in veterinary medicine and the sGnRH analog H-9205 in aquaculture.
Analog of salmon GnRH showing a very high affinity to GnRH receptors used in aquaculture. Usually applied in combination with a dopamine antagonist as domperidone or pimozide, sGnRHa induces ovulation and spawning in a large number of fish species.