Dog;Mouse;Pig;Rat Peptide YY (13-36) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4016635.0500 4016635.1000
H-9185.0500BAEA 326.8 USD
H-9185.0500BA H-9185.1000BA
Dog;Mouse;Pig;Rat Peptide YY (13-36) Trifluoroacetate
Proteins and Peptides
This C-terminal fragment was shown to suppress the noradrenaline release from sympathetic nerve endings. It thereby mimics the effects of PYY and NPY at presynaptic (Y₂) receptors. The peptide was also able to compete with NPY for essentially all binding sites in rat brain.