Human Peptide YY Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4013882.0500
4013882.1000
H-9180.0500BAEA
429.44
USD
H-9180.0500BA
H-9180.1000BA
Human Peptide YY Trifluoroacetate
Proteins and Peptides
Peptide YY has been isolated from human colonic extracts. It is identical in sequence to porcine PYY except for two amino acid replacements.