Palmitoyl-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-Ala-Gly-OH
Supplier: BACHEM AMERICAS INC
4018855.0005 4018855.0025
H-8820.0005BAEA 571.91 USD
H-8820.0005BA H-8820.0025BA
Palmitoyl-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-Ala-Gly-OH
Proteins and Peptides
See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.
This synthetic lipopeptide analog, corresponding to the N-terminal region of a bacterial lipoprotein, is a potent activator of macrophages. It is suggested that the uptake of this lipopeptide into macrophages involves the aggregation of membrane proteins.