SV40 Nuclear Transport Signal Peptide Analog, Bachem
Supplier: BACHEM AMERICAS INC
4026251.0001 4026251.0005
H-8120.0001BAEA 150.74 USD
H-8120.0001BA H-8120.0005BA
SV40 Nuclear Transport Signal Peptide Analog, Bachem
Proteins and Peptides
NLS, cell-penetrating peptide containing 7 amino acids homologous to the SV40 T antigen, was capable of inducing the transport of several carrier proteins to the nucleus. The terminal cysteine permits the attachment of this peptide to any protein using a heterobifunctional crosslinking reagent. Coupling to plasmid DNA has also been realized by M.G.Sebastyén and coworkers. Heterodimerization of TAT and NLS yields an even more effective transfecting peptide.