(Des-Gly¹⁰,D-Leu⁶,[¹³C₆]Leu⁷,Pro-NHEt⁹)-LHRH Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4048762.0500 4048762.1000
H-6258.0500BAEA 816.97 USD
H-6258.0500BA H-6258.1000BA
(Des-Gly¹⁰,D-Leu⁶,[¹³C₆]Leu⁷,Pro-NHEt⁹)-LHRH Trifluoroacetate
Proteins and Peptides

LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).


Heavy isotope-labeled leuprorelin useful for pharmacokinetic/pharmacodynamic studies in combination with Bachem product H-4060 (Leuprolide) or the corresponding generic API.

Order Now

SPECIFICATIONS

Protein/Peptide Name (Des-Gly¹⁰,D-Leu⁶,[¹³C₆]Leu⁷,Pro-NHEt⁹)-LHRH Trifluoroacetate
Protein Synonyms ([¹³C₆]Leu7)-Leuprolide
MDL No. MFCD18782382
Sequence Pyr-His-Trp-Ser-Tyr-D-Leu-[¹³C₆]Leu-Arg-Pro-NHEt
Molecular Weight 1215.35
Molecular Formula C₅₃¹³C₆H₈₄N₁₆O₁₂
Storage Conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR