E. coli Enterotoxin STp Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4044297.0500 4044297.1000
H-6248.0500BAEA 921.72 USD
H-6248.0500BA H-6248.1000BA
E. coli Enterotoxin STp Trifluoroacetate
Proteins and Peptides
Enterotoxigenic E.coli are able to produce several toxic peptides which may cause acute diarrhea in humans and domestic animals. The E.coli heat-stable enterotoxin STp is syntheszied in vivo as a 72 amino acid precursor consisting of pre-, pro-, and mature region. Mature STp is composed of 18 amino acids containing three intramolecular disulfide bonds, which seem to be important for the correct conformation of the biologically active structure.