Human Neuromedin S Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4048153.0500 4048153.1000
H-6232.0500BAEA 571.91 USD
H-6232.0500BA H-6232.1000BA
Human Neuromedin S Trifluoroacetate
Proteins and Peptides
The 33-amino acid neuropeptide, neuromedin S (NMS), was originally isolated from rat brain as an endogenous ligand for two orphan G protein-coupled receptors FM-3/GPR66 and FM-4/TGR-1, which have been identified as neuromedin U (NMU) receptors. hNMS-33 is specifically expressed in the suprachiasmatic nuclei (SCN) of the hypothalamus. It has been shown that NMS is implicated in the regulation of circadian rhythms through autocrine and/or paracrine actions.