Human GRPP Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4044528.0500 4044528.1000
H-6062.0500BAEA 490.19 USD
H-6062.0500BA H-6062.1000BA
Human GRPP Trifluoroacetate
Proteins and Peptides
For the long-acting GLP-1 analog liraglutide see H-6724.
GRPP (human) also known as glicentin-related polypeptide (human) is a synthetic analog of a cleavage product resulting from proglucagon processing in pancreatic α- and intestinal L-cells.