H-p-Chloro-Phe-D-Cys-beta-(3-pyridyl)-Ala-D-Trp-N-Me-Lys-Thr-Cys-2-Nal-NH₂ Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4041641.0500
4041641.1000
H-5884.0500BAEA
441.71
USD
H-5884.0500BA
H-5884.1000BA
H-p-Chloro-Phe-D-Cys-beta-(3-pyridyl)-Ala-D-Trp-N-Me-Lys-Thr-Cys-2-Nal-NH₂ Trifluoroacetate
Proteins and Peptides
This synthetic peptide analog of somatostatin-14 had a high affinity of 5.98 nM for the somatostatin receptor subtype 5 (sst5) (compared to 1.4 nM for somatostatin-14). Therefore, it represents the first antagonist with high affinity for sst5.