beta-Amyloid (31-35)
Supplier: BACHEM AMERICAS INC
4041418.0001 4041418.0005
H-5866.0001BAEA 111.72 USD
H-5866.0001BA H-5866.0005BA
beta-Amyloid (31-35)
Proteins and Peptides
The neurotoxicity of the short fragment IIGLM is similar to that of amyloid β-protein (1-42) (H-1368). Aβ 31-35 induced apoptosis in undifferentiated PC 12 cells.