BQ-610
Supplier: BACHEM AMERICAS INC
4034542.0500 4034542.1000
H-4914.0500BAEA 147.03 USD
H-4914.0500BA H-4914.1000BA
BQ-610
Proteins and Peptides
Endothelins are peptides with exceptional vasoconstrictor potency. They play an important role in intercellular communications.
BQ-610 is a very potent ET-A receptor antagonist which is about 30000 times more selective for ET-A receptors than for ET-B receptors. Furthermore, it has been found that BQ-610 strongly antagonizes ET-1 induced concentration in porcine coronary artery with a pA₂ value of 8.2.