KALA Amphipathic Peptide
Supplier: BACHEM AMERICAS INC
4030798.0500 4030798.1000
H-4096.0500BAEA 159.86 USD
H-4096.0500BA H-4096.1000BA
KALA Amphipathic Peptide
Proteins and Peptides
KALA amphipathic peptide, a low-molecular weight cationic amphipathic peptide was able to bind to DNA, to destabilize membranes and to mediate transfection of plasmid DNA in various cell lines. Thus, KALA provides a starting point for a family of peptides that incorporate other functions to improve DNA delivery systems. For further cell-permeable peptides see the product family 'Antennapedia Homeobox Peptides', and our HIV-tat fragments.