Human;Mouse;Rat (Asn¹⁰,Leu¹¹,D-Trp¹²)-pTH-Related Protein (7-34) amide
Supplier: BACHEM AMERICAS INC
4031198.0500 4031198.1000
H-3274.0500BAEA 552.66 USD
H-3274.0500BA H-3274.1000BA
Human;Mouse;Rat (Asn¹⁰,Leu¹¹,D-Trp¹²)-pTH-Related Protein (7-34) amide
Proteins and Peptides
This pTHrP (7-34) amide analog was found to be an effective antagonist of pTH-stimulated cAMP production in ROS cells and was shown not to display partial agonism.