H-Glu(Met-OH)-OH
Supplier: BACHEM AMERICAS INC
4000709.0250
4000709.1000
H-3195.0250BAEA
361.17
USD
H-3195.0250BA
H-3195.1000BA
H-Glu(Met-OH)-OH
Proteins and Peptides
See also G-4745, H-3996, the branched substrates H-2416, and L-2070, and the product families 'Glutathione-Related Peptides' and 'Ophthalmic Acid'.
γ-Glu-Met and further kokumi-active γ-glutamyl dipeptides have been isolated from mature Gouda cheese. These branched dipeptides are responsible for the complex taste of the cheese.